siRNA Details
Gene Symbol: Sepn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029100.2 | NP_083376.2 | 10 | 1419 | ||
GenBank mRNA | BC043665.1 | - | 2 | 107 | ||
BU057838.1 | - | 2 | 106 | |||
CF737950.1 | - | 4 | 461 |
