siRNA Details
Gene Symbol: Meikin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB987828.1 | - | 4 | 336 | ||
AK015073.1 | - | 4 | 354 | |||
BC107368.1 | - | 3 | 444 | 40056257 | ||
BC107369.1 | - | 4 | 353 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Meikin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB987828.1 | - | 4 | 336 | ||
AK015073.1 | - | 4 | 354 | |||
BC107368.1 | - | 3 | 444 | 40056257 | ||
BC107369.1 | - | 4 | 353 |