siRNA Details
Gene Symbol: 4930402F06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080709.2 | NP_001074178.1 | 6 | 1192 | ||
NM_001272040.1 | NP_001258969.1 | 5 | 1108 | |||
GenBank mRNA | AK015047.1 | - | 6 | 1191 | ||
BC115676.1 | - | 5 | 991 | 40104161 | ||
BC115677.1 | - | 5 | 991 | 40104162 |
