siRNA Details
Gene Symbol: Morn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029112.1 | NP_083388.1 | 1 | 122 | ||
GenBank mRNA | AK015339.1 | - | 1 | 275 | ||
AK077141.1 | - | 1 | 319 | |||
BC019987.1 | - | 1 | 122 | 4458628 |
