siRNA Details
Gene Symbol: 4930502E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029142.1 | NP_083418.1 | 1, 2 | 226 | ||
GenBank mRNA | AK015678.1 | - | 1, 2 | 232 | ||
BC115663.1 | - | 1, 2 | 226 | 40103303 | ||
BC115664.1 | - | 1, 2 | 226 | 40103304 |
