siRNA Details
Gene Symbol: Kif27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175214.3 | NP_780423.2 | 14, 15 | 3365 | ||
GenBank mRNA | AK032912.1 | - | 7, 8 | 1220 | ||
BC116646.1 | - | 12, 13 | 2204 | 40086396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
