siRNA Details
Gene Symbol: Zcchc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029158.2 | NP_083434.1 | 1 | 221 | ||
GenBank mRNA | AK015789.1 | - | 1 | 223 | ||
BC145772.1 | - | 1 | 210 | 40131075 | ||
BC145774.1 | - | 1 | 210 | 40131077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
