siRNA Details
Gene Symbol: Uhrf1bp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029166.2 | NP_083442.2 | 6 | 879 | ||
GenBank mRNA | AB093261.1 | - | 6 | 844 | ||
AK033300.1 | - | 7 | 887 | |||
AK087280.1 | - | 4 | 431 | |||
AK141050.1 | - | 6 | 888 | |||
BB529788.2 | - | 2 | 271 | |||
BC030383.1 | - | 838 | ||||
BC132134.1 | - | 6 | 672 | 40130411 | ||
BC138092.1 | - | 6 | 672 | 8861111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
