siRNA Details
Gene Symbol: Meiob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029197.1 | NP_083473.1 | 12 | 1111 | ||
GenBank mRNA | AK015924.1 | - | 4 | 352 | ||
BC119085.1 | - | 4 | 410 | 8733834 | ||
BC119087.1 | - | 4 | 410 | 8733836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
