siRNA Details
Gene Symbol: 4930538K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029198.3 | NP_083474.1 | 6 | 701 | ||
GenBank mRNA | AK015991.1 | - | 6 | 659 | ||
AK053970.1 | - | 6 | 702 | |||
BC048569.1 | - | 6 | 652 | 6705687 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
