siRNA Details
Gene Symbol: Dusp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173745.5 | NP_776106.1 | 2 | 411 | ||
GenBank mRNA | AK015917.1 | - | 2 | 395 | ||
AK079664.1 | - | 2 | 407 | |||
AK081916.1 | - | 2 | 370 | |||
BC020036.1 | - | 2 | 405 | 3990751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
