siRNA Details
Gene Symbol: Rnf19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029219.1 | NP_083495.1 | 8, 9 | 1736 | ||
GenBank mRNA | AK015966.1 | - | 3, 4 | 420 | ||
AK183889.1 | - | 2, 3 | 261 | |||
BC052529.1 | - | 2, 3 | 338 | |||
BC117807.1 | - | 8, 9 | 1294 | |||
BC139808.1 | - | 8, 9 | 1297 |

More Information
Other Species Targeted

Gene Ontology Categories:
