siRNA Details
Gene Symbol: 4930564B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029230.1 | NP_083506.1 | 4, 5 | 627 | ||
GenBank mRNA | AK016216.1 | - | 4, 5 | 628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
