siRNA Details
Gene Symbol: Tmco5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029232.2 | NP_083508.1 | 6, 7 | 563 | ||
GenBank mRNA | AK016214.1 | - | 6, 7 | 554 | ||
BC050803.1 | - | 6, 7 | 563 | 6771315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
