siRNA Details
Gene Symbol: Ppp1r1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290743.1 | NP_001277672.1 | 2 | 506 | ||
NM_001290744.1 | NP_001277673.1 | 2 | 506 | |||
NM_172420.4 | NP_766008.2 | 2 | 506 | |||
GenBank mRNA | AK051670.1 | - | 2 | 507 | ||
AK083940.1 | - | 2 | 220 | |||
AK157871.1 | - | 2 | 365 | |||
BC120706.1 | - | 2 | 468 | 40129629 | ||
BC132149.1 | - | 2 | 468 | 40130426 |

More Information
Other Species Targeted

Gene Ontology Categories:
