siRNA Details
Gene Symbol: Fgfr1op | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197046.1 | NP_001183975.1 | 5 | 464 | ||
NM_201230.5 | NP_957682.1 | 5 | 464 | |||
GenBank mRNA | AK016110.1 | - | 5 | 467 | ||
AK028778.1 | - | 5 | 393 | |||
AK029470.1 | - | 5 | 463 | |||
AK031261.1 | - | 5 | 447 | |||
AK050389.1 | - | 5 | 402 | |||
BB615739.1 | - | 5 | 466 | |||
BC050774.1 | - | 5 | 416 | |||
BC058804.1 | - | 5 | 399 | |||
BC080717.1 | - | 5 | 451 | 4935625 | ||
BC108331.1 | - | 5 | 462 | 30434232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
