siRNA Details
Gene Symbol: Asxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270988.1 | NP_001257917.1 | 12 | 3713 | ||
NM_172421.5 | NP_766009.2 | 11 | 3569 | |||
GenBank mRNA | AK036839.1 | - | 11 | 3526 | ||
AK122540.1 | - | 3 | 2342 | |||
AK162455.1 | - | 3 | 2310 | |||
BC141140.1 | - | 12 | 3547 | 9055752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
