siRNA Details
Gene Symbol: 4930563D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029252.2 | NP_083528.1 | 1 | 714 | ||
GenBank mRNA | AK029653.1 | - | 1 | 682 | ||
BC147584.1 | - | 1 | 695 | 9056574 | ||
BC147622.1 | - | 1 | 695 | 9056612 |
