siRNA Details
Gene Symbol: 4930553J12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016107.1 | - | 490 | |||
BC062172.1 | - | 1 | 364 | |||
BC080822.1 | - | 1 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
