siRNA Details
Gene Symbol: Mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029280.4 | NP_083556.2 | 3 | 617 | ||
GenBank mRNA | AK013063.1 | - | 3 | 600 | ||
AK133594.1 | - | 3 | 607 | |||
BC027547.1 | - | 3 | 550 | 1531973 | ||
CJ063020.1 | - | 3 | 607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
