siRNA Details
Gene Symbol: Zfp820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029281.2 | NP_083557.2 | 5 | 983 | ||
GenBank mRNA | BC019219.1 | - | 5 | 1021 | 5066012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
