siRNA Details
Gene Symbol: Igfbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018741.2 | NP_061211.1 | 4 | 795 | ||
GenBank mRNA | AB006141.1 | - | 4 | 783 | ||
AK012705.1 | - | 4 | 797 | |||
AK013339.1 | - | 3 | 321 | |||
BC051960.1 | - | 4 | 765 | 5710646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
