siRNA Details
Gene Symbol: 1700008F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168369.1 | NP_001161841.1 | 10 | 993 | ||
NM_029292.3 | NP_083568.2 | 6 | 488 | |||
GenBank mRNA | AK005764.1 | - | 6 | 489 | ||
AK076548.1 | - | 13 | 1266 | |||
BC050962.1 | - | 9 | 515 | |||
BC061058.1 | - | 7 | 392 | |||
BC147560.1 | - | 10 | 998 | 9056550 |
Gene Symbol: Zfp821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU234026.1 | - | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
