siRNA Details
Gene Symbol: Phpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029293.2 | NP_083569.1 | 1 | 99 | ||
GenBank mRNA | AK005756.1 | - | 1 | 99 | ||
AK154580.1 | - | 1 | 120 | |||
BC028657.1 | - | 1 | 72 | 1246740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
