siRNA Details
Gene Symbol: 1700001C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172091.1 | NP_001165562.1 | 2 | 667 | ||
NM_029296.2 | NP_083572.1 | 4 | 949 | |||
GenBank mRNA | AK005579.1 | - | 2 | 668 | ||
BC028263.1 | - | 4 | 912 | 5124927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
