siRNA Details
Gene Symbol: Dynlrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029297.1 | NP_083573.1 | 3 | 238 | ||
GenBank mRNA | AK005789.1 | - | 3 | 238 | ||
BC048623.1 | - | 3 | 279 | 6743790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
