siRNA Details
Gene Symbol: Fabp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029310.1 | NP_083586.1 | 4 | 468 | ||
GenBank mRNA | AK005765.1 | - | 3 | 314 | ||
BC030910.1 | - | 4 | 468 | 4505673 | ||
EU733649.1 | - | 3 | 444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
