siRNA Details
Gene Symbol: Nhej1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029342.4 | NP_083618.3 | 412 | |||
GenBank mRNA | AK006481.1 | - | 406 | |||
BC006063.1 | - | 386 | 3591824 | |||
BC107252.1 | - | 331 | 40052353 | |||
BC107253.1 | - | 331 | 40052354 | |||
BC132359.1 | - | 302 | 40130636 | |||
BC132361.1 | - | 302 | 40130638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
