siRNA Details
Gene Symbol: Dynap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029346.1 | NP_083622.1 | 2 | 279 | ||
GenBank mRNA | AK009098.1 | - | 2 | 94 | ||
HQ404307.1 | - | 2 | 192 | |||
HQ404309.1 | - | 2 | 192 | |||
HQ404310.1 | - | 2 | 192 | |||
HQ404311.1 | - | 2 | 192 |
Gene Symbol: Astl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HQ404308.1 | - | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
