siRNA Details
Gene Symbol: Ndufaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127346.1 | NP_001120818.1 | 3 | 285 | ||
GenBank mRNA | AB183434.1 | - | 3 | 229 | ||
AK007894.1 | - | 3 | 276 | |||
AK166492.1 | - | 3 | 252 | |||
AY262012.1 | - | 3 | 257 | |||
BC048512.1 | - | 3 | 244 | |||
BC116975.1 | - | 3 | 251 | 40126294 | ||
BC116977.1 | - | 3 | 251 | 40126296 | ||
BY251841.1 | - | 3 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
