siRNA Details
Gene Symbol: Calml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138304.2 | NP_612177.1 | 3 | 219 | ||
GenBank mRNA | AK008039.1 | - | 3 | 205 | ||
AK146656.1 | - | 3 | 192 | |||
AY061807.1 | - | 3 | 219 | |||
BC013541.1 | - | 3 | 197 | 4241711 | ||
BC023475.1 | - | 3 | 184 | 5042406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
