siRNA Details
Gene Symbol: Gns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029364.3 | NP_083640.1 | 9, 10 | 1179 | ||
GenBank mRNA | AK030773.1 | - | 9, 10 | 1178 | ||
AK049162.1 | - | 9, 10 | 1180 | |||
AK054046.1 | - | 9, 10 | 1179 | |||
AK083597.1 | - | 9, 10 | 1157 | |||
AK149863.1 | - | 9, 10 | 1164 | |||
AK151037.1 | - | 7, 8 | 801 | |||
AK159562.1 | - | 9, 10 | 1146 | |||
AK165180.1 | - | 9, 10 | 1159 | |||
AK165917.1 | - | 9, 10 | 1129 | |||
AK169485.1 | - | 9, 10 | 1161 | |||
AK170791.1 | - | 9, 10 | 1175 | |||
AK195315.1 | - | 1, 2 | 32 | |||
AK195952.1 | - | 1, 2 | 32 | |||
AK197098.1 | - | 2, 3 | 147 | |||
AK198268.1 | - | 1, 2 | 32 | |||
AK198818.1 | - | 2, 3 | 147 | |||
AK201813.1 | - | 2, 3 | 149 | |||
AK205650.1 | - | 1, 2 | 32 | |||
AK208382.1 | - | 2, 3 | 147 | |||
AK213725.1 | - | 1, 2 | 32 | |||
BC055328.1 | - | 9, 10 | 1173 | 6400020 | ||
BC059047.1 | - | 1, 2 | 333 |
