siRNA Details
Gene Symbol: Tex30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029368.1 | NP_083644.1 | 7 | 758 | ||
GenBank mRNA | AK006487.1 | - | 5 | 790 | ||
AK160565.1 | - | 7 | 759 | |||
BC099557.1 | - | 6 | 671 | 30916402 | ||
BC116881.1 | - | 5 | 955 | 40126200 | ||
BC119032.1 | - | 5 | 1023 | 8733781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
