siRNA Details
Gene Symbol: Rasl10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145216.3 | NP_660251.1 | 1 | 255 | ||
GenBank mRNA | AF498251.1 | - | 1 | 77 | ||
AK044011.1 | - | 1 | 256 | |||
BC107200.1 | - | 2 | 136 | |||
BC107201.1 | - | 2 | 136 | 40051192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
