siRNA Details
Gene Symbol: Klf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029416.2 | NP_083692.2 | 3 | 1003 | ||
GenBank mRNA | AF508984.1 | - | 3 | 969 | ||
AK018361.1 | - | 3 | 969 | |||
AK139827.1 | - | 3 | 970 | |||
BC064748.1 | - | 3 | 947 | 30476790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
