siRNA Details
Gene Symbol: Klf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029416.2 | NP_083692.2 | 3 | 988 | ||
GenBank mRNA | AF508984.1 | - | 3 | 954 | ||
AK018361.1 | - | 3 | 954 | |||
AK139827.1 | - | 3 | 955 | |||
BC064748.1 | - | 3 | 932 | 30476790 |
