siRNA Details
Gene Symbol: Lppr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014763.1 | - | 2 | 433 | ||
AK032034.1 | - | 2 | 799 | |||
AK043629.1 | - | 2 | 423 | |||
AK082692.1 | - | 2 | 695 | |||
AY512657.1 | - | 2 | 346 | |||
BB611685.1 | - | 2 | 563 | |||
BC031879.1 | - | 2 | 663 | 4527789 |
