siRNA Details
Gene Symbol: Them4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029431.1 | NP_083707.1 | 3 | 430 | ||
GenBank mRNA | AK014834.1 | - | 3 | 363 | ||
AK038420.1 | - | 3 | 328 | |||
AK138397.1 | - | 3 | 430 | |||
BC022612.1 | - | 2 | 278 | |||
BC134396.1 | - | 3 | 426 | 40136254 |
