siRNA Details
Gene Symbol: 4930447C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029444.2 | NP_083720.1 | 7 | 606 | ||
GenBank mRNA | AK014811.1 | - | 7 | 619 | ||
AK015397.1 | - | 7 | 606 | |||
BC137875.1 | - | 8 | 681 | 8860894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
