siRNA Details
Gene Symbol: Hormad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029458.1 | NP_083734.1 | 9 | 787 | ||
GenBank mRNA | AK015939.1 | - | 10 | 789 | ||
BC120781.1 | - | 9 | 787 | 40129704 | ||
BC120783.1 | - | 9 | 787 | 40129706 | ||
BC144842.1 | - | 9 | 787 | 9053390 |
