siRNA Details
Gene Symbol: 4930578C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175228.3 | NP_780437.1 | 5 | 1199 | ||
GenBank mRNA | AK016298.1 | - | 5 | 1202 | ||
AK085770.1 | - | 5 | 1200 | |||
AK163928.1 | - | 7 | 1542 | |||
BC118515.1 | - | 5 | 1200 | 40105826 |
