siRNA Details
Gene Symbol: Samd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029489.3 | NP_083765.2 | 5 | 446 | ||
GenBank mRNA | AK044227.1 | - | 4 | 322 | ||
DV659349.1 | - | 5 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
