siRNA Details
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153567.3 | NP_705795.2 | 7 | 1657 | ||
GenBank mRNA | BC038019.1 | - | 5 | 862 | 3597423 | |
CK619668.1 | - | 3 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
