siRNA Details
Gene Symbol: Zc3h18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029993.1 | NP_001025164.1 | 8 | 1448 | ||
NM_001029994.1 | NP_001025165.1 | 7 | 1376 | |||
GenBank mRNA | AK031413.1 | - | 8 | 1397 | ||
AK131189.1 | - | 5 | 520 | |||
AK159881.1 | - | 8 | 1384 | |||
AK165989.1 | - | 6 | 727 | |||
BC030495.1 | - | 7 | 1356 | 5400494 | ||
BC094330.1 | - | 5 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
