siRNA Details
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029512.2 | NP_083788.2 | 3 | 587 | ||
NM_181734.3 | NP_859423.2 | 3 | 584 | |||
GenBank mRNA | AK033600.1 | - | 3 | 533 | ||
AK039450.1 | - | 3 | 585 | |||
AK043791.1 | - | 3 | 554 | |||
AK052794.1 | - | 3 | 550 | |||
AK081680.1 | - | 3 | 547 | |||
AK156641.1 | - | 3 | 550 | |||
BC052389.1 | - | 3 | 543 | 5718874 | ||
BC060726.1 | - | 2 | 453 | |||
CK640285.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
