siRNA Details
Gene Symbol: Gpsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029522.2 | NP_083798.2 | 8, 9 | 1463 | ||
GenBank mRNA | AK040996.1 | - | 8, 9 | 1352 | ||
AK143552.1 | - | 8, 9 | 1464 | |||
AY081187.1 | - | 8, 9 | 1307 | |||
BC021308.1 | - | 7, 8 | 1018 | 5007832 |
