siRNA Details
Gene Symbol: Faxc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175234.4 | NP_780443.2 | 3 | 753 | ||
GenBank mRNA | AK018081.1 | - | 3 | 722 | ||
AK082464.1 | - | 3 | 747 | |||
AK144924.1 | - | 3 | 754 | |||
BC059864.1 | - | 3 | 267 |

More Information
Other Species Targeted

Gene Ontology Categories:
