siRNA Details
Gene Symbol: Mcur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081059.3 | NP_001074528.1 | 4 | 668 | ||
GenBank mRNA | AK018089.1 | - | 4 | 639 | ||
AK041849.1 | - | 4 | 669 | |||
AK210508.1 | - | 3 | 177 | |||
AK216207.1 | - | 3 | 177 | |||
AK219679.1 | - | 3 | 177 | |||
BC113195.1 | - | 5 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
