siRNA Details
Gene Symbol: Abca6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147218.2 | NP_671751.2 | 35, 36 | 4723 | ||
GenBank mRNA | AF498361.1 | - | 35, 36 | 4675 | ||
BC132417.1 | - | 35, 36 | 4560 | 40130694 | ||
BC132419.1 | - | 35, 36 | 4560 | 40130696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
