siRNA Details
Gene Symbol: Abca6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166556.1 | NP_001160028.1 | 6 | 865 | ||
NM_001166557.1 | NP_001160029.1 | 6 | 865 | |||
NM_147218.2 | NP_671751.2 | 6 | 865 | |||
GenBank mRNA | AF498361.1 | - | 6 | 817 | ||
AK018242.1 | - | 6 | 850 | |||
AK040652.1 | - | 6 | 865 | |||
AK042934.1 | - | 6 | 837 | |||
AK053384.1 | - | 6 | 792 | |||
BC132417.1 | - | 6 | 702 | 40130694 | ||
BC132419.1 | - | 6 | 702 | 40130696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
